Product Details

SNP ID
rs74787412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53606484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGACGGCCACAGCATATCTGATG[A/G]GGGCCCATCCAATGTGGACAAAAGC
Phenotype
MIM: 611165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPRX PubMed Links

Gene Details

Gene
DPRX
Gene Name
divergent-paired related homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012728.1 Intron NP_001012746.1
XM_011527011.2 Intron XP_011525313.1
XM_011527012.2 Intron XP_011525314.1
Gene
LOC284379
Gene Name
solute carrier family 7 member 3 pseudogene
There are no transcripts associated with this gene.

View Full Product Details