Product Details

SNP ID
rs75639038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4477203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTCCAGGATCCGCAGACGTTT[C/T]GGTGGTGACACCTGCCACGCACACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDGFRP2 PubMed Links

Gene Details

Gene
HDGFRP2
Gene Name
hepatoma-derived growth factor-related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001520.2 Intron NP_001001520.1
NM_032631.3 Intron NP_116020.1
XM_011528375.2 Intron XP_011526677.1
XM_011528376.2 Intron XP_011526678.1
XM_011528377.2 Intron XP_011526679.1

View Full Product Details