Product Details

SNP ID
rs75136459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5725127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGATGGTCTGAGTTTTTATCGA[G/T]TGTTCTGTTTACTTCTCTCTCAGAT
Phenotype
MIM: 605490
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152784.3 Intron NP_689997.3
XM_011527885.2 Intron XP_011526187.1
XM_011527886.2 Intron XP_011526188.1
XM_011527887.2 Intron XP_011526189.1
XM_011527888.2 Intron XP_011526190.1
XM_011527889.2 Intron XP_011526191.1
XM_011527890.2 Intron XP_011526192.1
XM_011527891.1 Intron XP_011526193.1
XM_011527892.1 Intron XP_011526194.1
XM_017026564.1 Intron XP_016882053.1
XM_017026565.1 Intron XP_016882054.1
XM_017026566.1 Intron XP_016882055.1
Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details