Product Details

SNP ID
rs76527006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6141562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGAATGACTGGAACCCCAAAG[A/G]CTCAAGAAGGAGCTAAAGATCTTGA
Phenotype
MIM: 614363
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACSBG2 PubMed Links

Gene Details

Gene
ACSBG2
Gene Name
acyl-CoA synthetase bubblegum family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289177.1 167 Missense Mutation ACT,GCT T7A NP_001276106.1
NM_001289178.1 167 Missense Mutation ACT,GCT T7A NP_001276107.1
NM_001289179.1 167 Missense Mutation ACT,GCT T7A NP_001276108.1
NM_001289180.1 167 UTR 5 NP_001276109.1
NM_001321384.1 167 UTR 5 NP_001308313.1
NM_030924.4 167 Missense Mutation ACT,GCT T7A NP_112186.3
XM_005259653.3 167 Missense Mutation ACT,GCT T7A XP_005259710.1
XM_011528329.1 167 Intron XP_011526631.1
XM_017027332.1 167 Missense Mutation ACT,GCT T7A XP_016882821.1
XM_017027333.1 167 Missense Mutation ACT,GCT T7A XP_016882822.1
XM_017027334.1 167 Missense Mutation ACT,GCT T7A XP_016882823.1
Gene
LOC105372255
Gene Name
uncharacterized LOC105372255
There are no transcripts associated with this gene.

View Full Product Details