Product Details

SNP ID
rs75846021
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19514743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCTCGAGGCCTTCGGGATAGA[C/G]CACGCCGCGTTTCTGGCGCCGGGGC
Phenotype
MIM: 614997 MIM: 609435 MIM: 610712
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GATAD2A PubMed Links
Additional Information
For this assay, SNP(s) [rs7250893] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GATAD2A
Gene Name
GATA zinc finger domain containing 2A
There are no transcripts associated with this gene.

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
There are no transcripts associated with this gene.

Gene
TSSK6
Gene Name
testis specific serine kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032037.3 918 Missense Mutation CTC,GTC L229V NP_114426.1

View Full Product Details