Product Details

SNP ID
rs76643934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11164514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGGTCACGCATTCCTGGGCCA[A/G]GGAGTTGCTTCTAAGAGCTTAAAAT
Phenotype
MIM: 614610 MIM: 609394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KANK2 PubMed Links

Gene Details

Gene
KANK2
Gene Name
KN motif and ankyrin repeat domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136191.2 4961 UTR 3 NP_001129663.1
NM_015493.6 4961 UTR 3 NP_056308.3
Gene
SPC24
Gene Name
SPC24, NDC80 kinetochore complex component
There are no transcripts associated with this gene.

View Full Product Details