Product Details

SNP ID
rs79821860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9093743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATCCTCCTGATGGCCCGTCTC[A/G]TTTTCTGCGGCAGCCATGAGGTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004456.1 499 Missense Mutation ATT,GTT I167V NP_001004456.1
Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
There are no transcripts associated with this gene.

View Full Product Details