Product Details
- SNP ID
-
rs76142062
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15673617 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCTGCTGGTTGTGGGCTCCTGG[A/C]TACTCGCCCGCATCCTGGCTTGGAC
- Phenotype
-
MIM: 611485
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CYP4F12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16995378] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYP4F12
- Gene Name
- cytochrome P450 family 4 subfamily F member 12
View Full Product Details