Product Details

SNP ID
rs80017107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17441279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAGCCCTCCATCAGCCTCTCCA[A/G]CATACAGTTGGCACCATTCTTAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM221 PubMed Links

Gene Details

Gene
TMEM221
Gene Name
transmembrane protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190844.1 Intron NP_001177773.1
XM_011527603.2 Intron XP_011525905.1

View Full Product Details