Product Details
- SNP ID
-
rs75297459
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:59581121 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAGGAGGATCGCAGAAGGACATG[A/G]GTACTGGGGGTCACAAACGCCAGCA
- Phenotype
-
MIM: 608725
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PHACTR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34169680] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHACTR3
- Gene Name
- phosphatase and actin regulator 3
View Full Product Details