Product Details
- SNP ID
-
rs74340988
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:189557 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAATATCTCTTAGGATGTTATAG[C/T]ACATTTGGACAAGTTACCTGTGAGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB128
PubMed Links
Gene Details
- Gene
- DEFB128
- Gene Name
- defensin beta 128
View Full Product Details