Product Details

SNP ID
rs80135188
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1475938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGAGAGACTGAGCCAGGGACT[C/G]AAAAGCAAGGAGGTCTTGAACAGGC
Phenotype
MIM: 606610
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
NSFL1C PubMed Links

Gene Details

Gene
NSFL1C
Gene Name
NSFL1 cofactor
There are no transcripts associated with this gene.

Gene
SIRPB2
Gene Name
signal regulatory protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122962.1 1486 UTR 3 NP_001116434.1
NM_001134836.1 1486 UTR 3 NP_001128308.1
XM_005260708.4 1486 Intron XP_005260765.1
XM_005260709.3 1486 Intron XP_005260766.1
XM_011529221.2 1486 Intron XP_011527523.1
XM_011529223.2 1486 Intron XP_011527525.1
XM_011529224.2 1486 Intron XP_011527526.1
XM_011529225.2 1486 Intron XP_011527527.1

View Full Product Details