Product Details

SNP ID
rs76441912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:762099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTGGTTCAAATCCCAGTTCTA[C/T]TGTTTAAGCTGAGACTCGCCTCTGA
Phenotype
MIM: 613350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC52A3 PubMed Links

Gene Details

Gene
SLC52A3
Gene Name
solute carrier family 52 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033409.3 Intron NP_212134.3
XM_005260655.3 Intron XP_005260712.1
XM_011529148.1 Intron XP_011527450.1

View Full Product Details