Product Details

SNP ID
rs80083448
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:26920930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATATACACCTGCTTCACACAGA[A/C]AGCTTTCTGTAAAAAAATACAAAAG
Phenotype
MIM: 605007
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ADAMTS5 PubMed Links

Gene Details

Gene
ADAMTS5
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007038.4 6645 UTR 3 NP_008969.2
XM_011529448.2 6645 UTR 3 XP_011527750.1

View Full Product Details