Product Details

SNP ID
rs76037294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37376191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAGGAGGGAAGAACACAGTCC[A/G]TGTGCCCTTTACTGAGAAAAATGAA
Phenotype
MIM: 600855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYRK1A PubMed Links

Gene Details

Gene
DYRK1A
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001396.3 Intron NP_001387.2
NM_101395.2 Intron NP_567824.1
NM_130436.2 Intron NP_569120.1
NM_130438.2 Intron NP_569122.1
XM_005260933.4 Intron XP_005260990.1
XM_006723976.3 Intron XP_006724039.1
XM_006723977.3 Intron XP_006724040.1
XM_006723978.3 Intron XP_006724041.1
XM_006723979.3 Intron XP_006724042.1
XM_011529482.2 Intron XP_011527784.1
XM_011529483.2 Intron XP_011527785.1
XM_011529484.2 Intron XP_011527786.1
XM_011529485.2 Intron XP_011527787.1
XM_017028284.1 Intron XP_016883773.1
XM_017028285.1 Intron XP_016883774.1
XM_017028286.1 Intron XP_016883775.1

View Full Product Details