Product Details

SNP ID
rs78368941
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36918704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTGTTGTATCTGTTTGGCTGC[A/G]CATGGAAAATCCCTAAGCAACAGAG
Phenotype
MIM: 138981
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CSF2RB PubMed Links

Gene Details

Gene
CSF2RB
Gene Name
colony stimulating factor 2 receptor beta common subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000395.2 Intron NP_000386.1
XM_005261340.3 Intron XP_005261397.1
XM_011529903.2 Intron XP_011528205.1
XM_011529904.2 Intron XP_011528206.1
XM_011529905.2 Intron XP_011528207.1

View Full Product Details