Product Details

SNP ID
rs80326013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:17760412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACTTCTCTCCACATCTAATCC[A/G]TCAGTAAGTCTTGCCAGTTCTACCT
Phenotype
MIM: 612449 MIM: 609387
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GEN1 PubMed Links

Gene Details

Gene
GEN1
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130009.2 Intron NP_001123481.2
NM_182625.4 Intron NP_872431.4
XM_005262613.4 Intron XP_005262670.1
XM_006712005.3 Intron XP_006712068.1
XM_011532820.2 Intron XP_011531122.1
XM_011532821.2 Intron XP_011531123.1
XM_011532822.2 Intron XP_011531124.1
Gene
SMC6
Gene Name
structural maintenance of chromosomes 6
There are no transcripts associated with this gene.

View Full Product Details