Product Details

SNP ID
rs75444457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176686413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGCTCAACTTCACAGTAAGAGG[C/T]TAATAAAATAGAAGAAATTTGGGAA
Phenotype
MIM: 601528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VEGFC PubMed Links

Gene Details

Gene
VEGFC
Gene Name
vascular endothelial growth factor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005429.4 Intron NP_005420.1

View Full Product Details