Product Details

SNP ID
rs76370652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88717959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGTTGCCCTGTTGTCATTTCC[A/G]GTCTCGCAAAGGCACAGTGAATATC
Phenotype
MIM: 613299 MIM: 613300 MIM: 605200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM13A PubMed Links

Gene Details

Gene
FAM13A
Gene Name
family with sequence similarity 13 member A
There are no transcripts associated with this gene.

Gene
FAM13A-AS1
Gene Name
FAM13A antisense RNA 1
There are no transcripts associated with this gene.

Gene
HERC3
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 3
There are no transcripts associated with this gene.

View Full Product Details