Product Details

SNP ID
rs75516903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105684176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGGCTAATCAGAAGCTCACAA[C/T]GGTACCAATGCCACTATATAAATAC
Phenotype
MIM: 611355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF38 PubMed Links

Gene Details

Gene
ARHGEF38
Gene Name
Rho guanine nucleotide exchange factor 38
There are no transcripts associated with this gene.

Gene
INTS12
Gene Name
integrator complex subunit 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142471.1 Intron NP_001135943.1
NM_020395.3 Intron NP_065128.2
XM_005263148.4 Intron XP_005263205.1
XM_011532143.2 Intron XP_011530445.1
XM_011532145.2 Intron XP_011530447.1

View Full Product Details