Product Details

SNP ID
rs75268512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:81033943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATTTTAATGGAAGATCAGGTG[G/T]TGTGCTGTGTATGGGGCAGCTGCTG
Phenotype
MIM: 112263
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BMP3 PubMed Links

Gene Details

Gene
BMP3
Gene Name
bone morphogenetic protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006714291.3 Intron XP_006714354.1

View Full Product Details