Product Details
- SNP ID
-
rs74639819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:155926070 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAGAAGAAACGGAATCTGCAATA[C/T]CTAAGGGAAAAAAAAGGAATTATTA
- Phenotype
-
MIM: 600550
MIM: 191070
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTSO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1543108] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTSO
- Gene Name
- cathepsin O
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001334.2 |
981 |
Missense Mutation |
GAT,GGT |
D311G |
NP_001325.1 |
- Gene
- TDO2
- Gene Name
- tryptophan 2,3-dioxygenase
There are no transcripts associated with this gene.
View Full Product Details