Product Details
- SNP ID
-
rs75149124
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:161385872 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGAAGGTGTCATCAGGTATTGAC[A/C]AAACAAGCCACTGTCCTGGATTTGC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FSTL5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17040982] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSTL5
- Gene Name
- follistatin like 5
View Full Product Details