Product Details
- SNP ID
-
rs78555651
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:163338206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGGTGATGGCCTAAAATGGTCCC[C/T]ATACTGTTTGCTTCTTCACAGCAGG
- Phenotype
-
MIM: 162641
MIM: 602001
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NPY1R
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35397712] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NPY1R
- Gene Name
- neuropeptide Y receptor Y1
- Gene
- NPY5R
- Gene Name
- neuropeptide Y receptor Y5
View Full Product Details