Product Details

SNP ID
rs75722914
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:9961570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTGGGGTAGGGATAAAGGGT[A/G]TGGGTACAAGCCCCAAGTGAATCAT
Phenotype
MIM: 608700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR5697 PubMed Links

Gene Details

Gene
MIR5697
Gene Name
microRNA 5697
There are no transcripts associated with this gene.

Gene
NMNAT1
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297778.1 Intron NP_001284707.1
NM_001297779.1 Intron NP_001284708.1
NM_022787.3 Intron NP_073624.2
XM_011541971.2 Intron XP_011540273.1
XM_017002107.1 Intron XP_016857596.1
XM_017002108.1 Intron XP_016857597.1

View Full Product Details