Product Details

SNP ID
rs78794750
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:11069157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGTAGCCCCTTCAAAGCAGAC[A/G]GACTCTCACAGCCCAAGCAGCCATA
Phenotype
MIM: 605960 MIM: 182891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001998.2 Intron NP_001001998.1
NM_002685.3 Intron NP_002676.1
XM_005263475.2 Intron XP_005263532.2
XM_017001494.1 Intron XP_016856983.1
XM_017001495.1 Intron XP_016856984.1
XM_017001496.1 Intron XP_016856985.1
XM_017001497.1 Intron XP_016856986.1
Gene
SRM
Gene Name
spermidine synthase
There are no transcripts associated with this gene.

View Full Product Details