Product Details

SNP ID
rs80254932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112979299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGGTCAGAACCTTGTGGCAG[G/T]TCCACGAAGTGACAGTGTGACCCCA
Phenotype
MIM: 605542
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL36G PubMed Links

Gene Details

Gene
IL36G
Gene Name
interleukin 36, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278568.1 213 Intron NP_001265497.1
NM_019618.3 213 Missense Mutation GGT,GTT G45V NP_062564.1

View Full Product Details