Product Details

SNP ID
rs80179025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:117817945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTGAGGAATATATTTTTAATTT[C/T]ACTAACCTTGAAGAAAACCTCTCTC
Phenotype
MIM: 606355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX18 PubMed Links

Gene Details

Gene
DDX18
Gene Name
DEAD-box helicase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006773.3 Intron NP_006764.3

View Full Product Details