Product Details

SNP ID
rs79706951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105341102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTTGATGGTTACTTTACTTACT[A/G]GTACAGACTATGTAGTGGTGTTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf49 PubMed Links

Gene Details

Gene
C2orf49
Gene Name
chromosome 2 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286537.1 Intron NP_001273466.1
NM_024093.2 Intron NP_076998.1
XM_017004892.1 Intron XP_016860381.1
Gene
LOC100506473
Gene Name
uncharacterized LOC100506473
There are no transcripts associated with this gene.

View Full Product Details