Product Details

SNP ID
rs78429992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98602209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTTCAATAAGATGCACACTCA[C/T]TTTAATAGCATACAAAATTACCCAC
Phenotype
MIM: 613920 MIM: 600916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COA5 PubMed Links

Gene Details

Gene
COA5
Gene Name
cytochrome c oxidase assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008215.2 Intron NP_001008216.1
Gene
INPP4A
Gene Name
inositol polyphosphate-4-phosphatase type I A
There are no transcripts associated with this gene.

Gene
UNC50
Gene Name
unc-50 inner nuclear membrane RNA binding protein
There are no transcripts associated with this gene.

View Full Product Details