Product Details

SNP ID
rs80282720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98613945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGCTGGGAATTGCAAGTGCAA[C/T]GGCATAGAGGTGTGAACAGCACACT
Phenotype
MIM: 613920 MIM: 604623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COA5 PubMed Links

Gene Details

Gene
COA5
Gene Name
cytochrome c oxidase assembly factor 5
There are no transcripts associated with this gene.

Gene
MGAT4A
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A
There are no transcripts associated with this gene.

Gene
UNC50
Gene Name
unc-50 inner nuclear membrane RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263913.4 Intron XP_005263970.1
XM_006712403.3 Intron XP_006712466.1
XM_006712404.2 Intron XP_006712467.1

View Full Product Details