Product Details

SNP ID
rs75227546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101992894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTCCTTGCCCATGACCTTCTT[C/T]ACCTGTGTGAAGGTGGCTGGAAGCA
Phenotype
MIM: 147811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1R2 PubMed Links

Gene Details

Gene
IL1R2
Gene Name
interleukin 1 receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261419.1 Intron NP_001248348.1
NM_004633.3 Intron NP_004624.1
XM_006712734.3 Intron XP_006712797.1
XM_006712736.3 Intron XP_006712799.1
XM_011511801.2 Intron XP_011510103.1
XM_011511803.2 Intron XP_011510105.1
XM_011511804.2 Intron XP_011510106.1
XM_011511805.2 Intron XP_011510107.1
XM_011511807.1 Intron XP_011510109.1
XM_011511808.2 Intron XP_011510110.1
XM_017004889.1 Intron XP_016860378.1
Gene
LINC01127
Gene Name
long intergenic non-protein coding RNA 1127
There are no transcripts associated with this gene.

View Full Product Details