Product Details

SNP ID
rs77815959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68134829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAACAAGAGGCATACAACTGGT[G/T]TATATGCTAAATGGTGCCCTGGAAG
Phenotype
MIM: 610729
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR92 PubMed Links

Gene Details

Gene
WDR92
Gene Name
WD repeat domain 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256476.1 Intron NP_001243405.1
NM_138458.3 Intron NP_612467.1

View Full Product Details