Product Details

SNP ID
rs76826407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25928040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGATGTTAATATCTTCGTTTAGC[C/G]AGGGGTAGAATGACATTGACTCCTC
Phenotype
MIM: 602845
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF3C PubMed Links

Gene Details

Gene
KIF3C
Gene Name
kinesin family member 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002254.6 3974 UTR 3 NP_002245.4
XM_005264299.2 3974 UTR 3 XP_005264356.2

View Full Product Details