Product Details

SNP ID
rs76341053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26035674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACCCATCCGCTCCGCAATAAAT[G/T]TCTTAAAAGACACATGGCTCTCAGC
Phenotype
MIM: 612672
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB10 PubMed Links

Gene Details

Gene
RAB10
Gene Name
RAB10, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016131.4 Intron NP_057215.3

View Full Product Details