Product Details

SNP ID
rs77370238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27782842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTTAAGGGTTGGGGAATTATT[C/T]CCTGAAGTTATGCTCTATCTCCTTA
Phenotype
MIM: 610059 MIM: 611132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL33 PubMed Links

Gene Details

Gene
MRPL33
Gene Name
mitochondrial ribosomal protein L33
There are no transcripts associated with this gene.

Gene
RBKS
Gene Name
ribokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287580.1 Intron NP_001274509.1
NM_022128.2 Intron NP_071411.1

View Full Product Details