Product Details

SNP ID
rs78307472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3133157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGTATGCAAGAATATGAGAGA[C/T]TAGAATAGTGTATTATTGTCATATC
Phenotype
MIM: 612907
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRNT1 PubMed Links

Gene Details

Gene
TRNT1
Gene Name
tRNA nucleotidyl transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302946.1 Intron NP_001289875.1
NM_182916.2 Intron NP_886552.2
XM_011533776.2 Intron XP_011532078.1
XM_011533777.2 Intron XP_011532079.1
XM_011533778.2 Intron XP_011532080.1

View Full Product Details