Product Details

SNP ID
rs74407198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3148898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATGTGTATTAGGTGTCATATA[C/T]AATGGTAATATGCCTGTCTTTAAAG
Phenotype
MIM: 609262 MIM: 612907
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRBN PubMed Links

Gene Details

Gene
CRBN
Gene Name
cereblon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173482.1 3181 Intron NP_001166953.1
NM_016302.3 3181 Intron NP_057386.2
XM_005265202.3 3181 Intron XP_005265259.1
XM_011533791.2 3181 UTR 3 XP_011532093.1
XM_011533793.2 3181 Intron XP_011532095.1
XM_011533794.2 3181 Intron XP_011532096.1
Gene
TRNT1
Gene Name
tRNA nucleotidyl transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302946.1 3181 UTR 3 NP_001289875.1
NM_182916.2 3181 UTR 3 NP_886552.2
XM_011533776.2 3181 UTR 3 XP_011532078.1
XM_011533777.2 3181 UTR 3 XP_011532079.1
XM_011533778.2 3181 Intron XP_011532080.1

View Full Product Details