Product Details

SNP ID
rs74919004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33150844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCATAAAAAAGATCAGGATTTC[A/C]CTCTACCACACAAAAACACTTAGAC
Phenotype
MIM: 605497
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
There are no transcripts associated with this gene.

Gene
SUSD5
Gene Name
sushi domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015551.1 4206 UTR 3 NP_056366.1
XM_005265034.3 4206 UTR 3 XP_005265091.1
XM_017006137.1 4206 UTR 3 XP_016861626.1

View Full Product Details