Product Details

SNP ID
rs74562335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36712836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACAATCTGTTTATCCATGCAC[C/T]AAATCATGCCCATTCAAGATGTTTC
Phenotype
MIM: 613167
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCLK3 PubMed Links

Gene Details

Gene
DCLK3
Gene Name
doublecortin like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033403.1 4889 UTR 3 NP_208382.1
XM_011534167.2 4889 UTR 3 XP_011532469.1
XM_011534168.2 4889 UTR 3 XP_011532470.1
XM_011534169.1 4889 Intron XP_011532471.1

View Full Product Details