Product Details
- SNP ID
-
rs75220201
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:74310494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAAAAAAAGAAGATAA[C/T]ATACTGCAGAAAATTAAAGAAGCAT
- Phenotype
-
MIM: 614047
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPGN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs532193046] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPGN
- Gene Name
- epithelial mitogen
- Gene
- LOC105377276
- Gene Name
- uncharacterized LOC105377276
There are no transcripts associated with this gene.
- Gene
- MTHFD2L
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
There are no transcripts associated with this gene.
View Full Product Details