Product Details

SNP ID
rs75813813
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176526701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACACGGGCAGCCAAAGGCAGC[A/G]TGGGCGGTCGTTGGTATACATCATA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RNF44 PubMed Links

Gene Details

Gene
RNF44
Gene Name
ring finger protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014901.4 4151 UTR 3 NP_055716.1
XM_005265840.2 4151 Intron XP_005265897.1
XM_005265841.4 4151 Intron XP_005265898.1
XM_005265842.4 4151 Intron XP_005265899.1
XM_005265843.2 4151 Intron XP_005265900.1
XM_005265844.3 4151 Intron XP_005265901.1
XM_005265845.4 4151 Intron XP_005265902.1
XM_006714831.2 4151 Intron XP_006714894.1
XM_006714832.2 4151 Intron XP_006714895.1
XM_011534466.2 4151 Intron XP_011532768.1
XM_011534467.2 4151 Intron XP_011532769.1

View Full Product Details