Product Details

SNP ID
rs77271640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180110222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCACAGCTCGCTGGGGTGGGAGC[A/G]GCTCAGACAGGTTTATTCATCAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGEF1C PubMed Links

Gene Details

Gene
RASGEF1C
Gene Name
RasGEF domain family member 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175062.3 Intron NP_778232.2

View Full Product Details