Product Details

SNP ID
rs76129633
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136799248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACACAAACGCCGCCCCTCCCCT[C/T]TGCCTGGCGAGCAGGGCAGGAGCTG
Phenotype
MIM: 615955 MIM: 610615
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC183 PubMed Links

Gene Details

Gene
CCDC183
Gene Name
coiled-coil domain containing 183
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039374.4 Intron NP_001034463.4
Gene
CCDC183-AS1
Gene Name
CCDC183 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RABL6
Gene Name
RAB, member RAS oncogene family-like 6
There are no transcripts associated with this gene.

Gene
TMEM141
Gene Name
transmembrane protein 141
There are no transcripts associated with this gene.

View Full Product Details