Product Details
- SNP ID
-
rs74613026
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:114281835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACCTCGCGAGGAGAAGAGGACG[G/T]CCCTGAGCAAGGTGGGGACGGGGGC
- Phenotype
-
MIM: 603461
MIM: 605530
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDC16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs556799572] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDC16
- Gene Name
- cell division cycle 16
There are no transcripts associated with this gene.
- Gene
- MIR4502
- Gene Name
- microRNA 4502
There are no transcripts associated with this gene.
- Gene
- UPF3A
- Gene Name
- UPF3 regulator of nonsense transcripts homolog A (yeast)
View Full Product Details