Product Details

SNP ID
rs79689562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19823741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAAACTGAGTAGTTTTACTTTA[A/T]TAATAATGTTGAGAATGCTTGTTCA
Phenotype
MIM: 616443
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZMYM5 PubMed Links

Gene Details

Gene
ZMYM5
Gene Name
zinc finger MYM-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039649.2 3370 Intron NP_001034738.1
NM_001039650.2 3370 Intron NP_001034739.1
NM_001142684.1 3370 UTR 3 NP_001136156.1
XM_005266593.3 3370 UTR 3 XP_005266650.1
XM_005266594.2 3370 UTR 3 XP_005266651.1
XM_006719894.2 3370 UTR 3 XP_006719957.1
XM_011535309.1 3370 UTR 3 XP_011533611.1
XM_011535310.1 3370 UTR 3 XP_011533612.1
XM_017020849.1 3370 UTR 3 XP_016876338.1
XM_017020850.1 3370 UTR 3 XP_016876339.1
XM_017020851.1 3370 Intron XP_016876340.1

View Full Product Details