Product Details

SNP ID
rs80298677
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:142752223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACATGGGGTGGAATGTTTTCCC[A/T]TATTTTTAGATGAGTATATAGATCA
Phenotype
MIM: 143054
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
HIVEP2 PubMed Links

Gene Details

Gene
HIVEP2
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006734.3 8933 UTR 3 NP_006725.3
XM_017010805.1 8933 UTR 3 XP_016866294.1

View Full Product Details