Product Details
- SNP ID
-
rs79981792
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:143848321 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCTTAAAAAACATTAAATTGTA[C/T]ACTTTAGGTGAATTGTATGTGAATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LTV1
PubMed Links
Gene Details
- Gene
- LTV1
- Gene Name
- LTV1 ribosome biogenesis factor
There are no transcripts associated with this gene.
View Full Product Details