Product Details

SNP ID
rs76838309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169726925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGACCACTTAGATTATGCAGA[G/T]ATACGTGAAAATAAGGCCCAAGACC
Phenotype
MIM: 613069 MIM: 186977
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF10 PubMed Links

Gene Details

Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
TCTE3
Gene Name
t-complex-associated-testis-expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006715554.3 Intron XP_006715617.1
XM_011536093.2 Intron XP_011534395.1

View Full Product Details