Product Details

SNP ID
rs79271379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20781163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAGAGTTTAATCCAGACTCTGC[A/G]CAAGAAGTTAATCTGCCAAACCCTA
Phenotype
MIM: 601981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNASE6 PubMed Links

Gene Details

Gene
RNASE6
Gene Name
ribonuclease A family member k6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005615.4 113 UTR 5 NP_005606.1
XM_017021566.1 113 Intron XP_016877055.1
XM_017021567.1 113 Intron XP_016877056.1

View Full Product Details